missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, 95%

Catalog No. AAH6201803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6201803 1 g
AAH6201806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6201803 Supplier Thermo Scientific Chemicals Supplier No. H6201803
Only null left

CAS: 31687-58-8 | C28H48N4O6 | 536.714 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31687-58-8
Molecular Formula C28H48N4O6
Molecular Weight (g/mol) 536.714
InChI Key WBGMQHNUPJENDC-MERQFXBCSA-N
Synonym boc-his boc-oh.dcha, boc-his boc-oh dcha, boc-his boc-ohdcha, boc-his boc .dcha, n,1-bis tert-butoxycarbonyl-l-histidine, compound with dicyclohexylamine 1:1, pubchem18934, n,1-bis boc-l-histidine dicyclohexylamine salt, boc-his boc-ohcha, boc-his boc-oh?cha, boc-his boc-oh toluene solvate
PubChem CID 16211319
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]imidazol-4-yl]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C(=O)OC(C)(C)C)C(=O)O.C1CCC(CC1)NC2CCCCC2

Specifications

Melting Point 160°C to 165°C (decomposition)
Color White
Quantity 1 g
Formula Weight 536.7
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N,N'-Di-Boc-L-histidine dicyclohexylammonium salt
EINECSNumber 250-764-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only