missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di(4-biphenylyl)-N,N'-diphenylbenzidine 98.0+%, TCI America™

Catalog No. d49285g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
D49285G 5 g
D49281G 1 g
2 options

Catalog No. D49285G

Supplier: TCI America D49285G

Only null left

Chemical Identifiers

CAS 134008-76-7
Molecular Formula C48H36N2
Molecular Weight (g/mol) 640.83
InChI Key SDVOZSYGHFDAKX-UHFFFAOYSA-N
PubChem CID 22251228
IUPAC Name N,4-diphenyl-N-[4-[4-(N-(4-phenylphenyl)anilino)phenyl]phenyl]aniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N(C3=CC=CC=C3)C4=CC=C(C=C4)C5=CC=C(C=C5)N(C6=CC=CC=C6)C7=CC=C(C=C7)C8=CC=CC=C8

Specifications

Melting Point 213°C
Color White-Yellow
Quantity 5 g
Formula Weight 640.83
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Di(4-biphenylyl)-N,N′-diphenylbenzidine