missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Di-2-naphthyl-1,4-phenylenediamine 98.0+%, TCI America™
$57.79 - $328.08
Chemical Identifiers
| CAS | 93-46-9 |
|---|---|
| Molecular Formula | C26H20N2 |
| Molecular Weight (g/mol) | 360.46 |
| MDL Number | MFCD00053487 |
| InChI Key | VETPHHXZEJAYOB-UHFFFAOYSA-N |
| PubChem CID | 7142 |
| IUPAC Name | 1-N,4-N-dinaphthalen-2-ylbenzene-1,4-diamine |
| SMILES | C1=CC=C2C=C(C=CC2=C1)NC3=CC=C(C=C3)NC4=CC5=CC=CC=C5C=C4 |
Chemical Identifiers
| 93-46-9 | |
| 360.46 | |
| VETPHHXZEJAYOB-UHFFFAOYSA-N | |
| 1-N,4-N-dinaphthalen-2-ylbenzene-1,4-diamine |
| C26H20N2 | |
| MFCD00053487 | |
| 7142 | |
| C1=CC=C2C=C(C=CC2=C1)NC3=CC=C(C=C3)NC4=CC5=CC=CC=C5C=C4 |
Specifications
| 93-46-9 | |
| MFCD00053487 | |
| VETPHHXZEJAYOB-UHFFFAOYSA-N | |
| 1-N,4-N-dinaphthalen-2-ylbenzene-1,4-diamine | |
| 7142 | |
| ≥98.0% (HPLC) | |
| N,N′-Di-2-naphthyl-1,4-phenylenediamine |
| C26H20N2 | |
| 25 g | |
| C1=CC=C2C=C(C=CC2=C1)NC3=CC=C(C=C3)NC4=CC5=CC=CC=C5C=C4 | |
| 360.46 | |
| 360.46 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0346
RTECSNumber : ST2100000
TSCA : Yes