missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Di-2-naphthyl-1,4-phenylenediamine 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 500 g |
| Formula Weight | 360.46 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N,N′-Di-2-naphthyl-1,4-phenylenediamine |
Safety and Handling
| EINECSNumber | (4)-0346 |
| RTECSNumber | ST2100000 |
| TSCA | Yes |