missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-2-naphthyl-1,4-phenylenediamine 98.0+%, TCI America™

Catalog No. D081225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D081225G 25 g
D0812500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D081225G Supplier TCI America Supplier No. D081225G
Only null left

Chemical Identifiers

CAS 93-46-9
Molecular Formula C26H20N2
Molecular Weight (g/mol) 360.46
MDL Number MFCD00053487
InChI Key VETPHHXZEJAYOB-UHFFFAOYSA-N
PubChem CID 7142
IUPAC Name 1-N,4-N-dinaphthalen-2-ylbenzene-1,4-diamine
SMILES C1=CC=C2C=C(C=CC2=C1)NC3=CC=C(C=C3)NC4=CC5=CC=CC=C5C=C4

Specifications

Quantity 25 g
Formula Weight 360.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Di-2-naphthyl-1,4-phenylenediamine
EINECSNumber (4)-0346
RTECSNumber ST2100000
TSCA Yes