missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine (purified by sublimation) 98.0+%, TCI America™

Catalog No. D39701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39701G 1 g
D39705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39701G Supplier TCI America Supplier No. D39701G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123847-85-8
Molecular Formula C44H32N2
Molecular Weight (g/mol) 588.754
MDL Number MFCD03093246
InChI Key IBHBKWKFFTZAHE-UHFFFAOYSA-N
Synonym n4,n4'-di naphthalen-1-yl-n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, alpha-npb, n,n'-bis naphthalen-1-yl-n,n'-bis phenyl benzidine, 4,4'-bis n-1-naphthyl-n-phenylamino biphenyl, n,n'-di 1-naphthyl-n,n'-diphenylbenzidine, n,n'-bis-1-naphthalenyl-n,n'-bis-phenyl-1,1'-biphenyl-4,4'-diamine, 1,1'-biphenyl-4,4'-diamine, n,n'-di-1-naphthalenyl-n,n'-diphenyl, n,n'-di-1-naphthyl-n,n'-diphenylbenzidine
PubChem CID 5069127
IUPAC Name N-[4-[4-(N-naphthalen-1-ylanilino)phenyl]phenyl]-N-phenylnaphthalen-1-amine
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC6=CC=CC=C65)C7=CC=CC8=CC=CC=C87

Specifications

Melting Point 281°C
Quantity 1 g
Formula Weight 588.75
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Di-1-naphthyl-N,N′-diphenylbenzidine (purified by sublimation)