missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di(1-naphthyl)-N,N'-diphenylbenzidine, 98%, Thermo Scientific Chemicals

Catalog No. p-7055826
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64502MD 250 mg
AAH6450203 1 g
AAH6450206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64502MD Supplier Thermo Scientific Chemicals Supplier No. H64502MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123847-85-8
Molecular Formula C44H32N2
Molecular Weight (g/mol) 588.754
MDL Number MFCD03093246
InChI Key IBHBKWKFFTZAHE-UHFFFAOYSA-N
Synonym n4,n4'-di naphthalen-1-yl-n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, alpha-npb, n,n'-bis naphthalen-1-yl-n,n'-bis phenyl benzidine, 4,4'-bis n-1-naphthyl-n-phenylamino biphenyl, n,n'-di 1-naphthyl-n,n'-diphenylbenzidine, n,n'-bis-1-naphthalenyl-n,n'-bis-phenyl-1,1'-biphenyl-4,4'-diamine, 1,1'-biphenyl-4,4'-diamine, n,n'-di-1-naphthalenyl-n,n'-diphenyl, n,n'-di-1-naphthyl-n,n'-diphenylbenzidine
PubChem CID 5069127
IUPAC Name N-[4-[4-(N-naphthalen-1-ylanilino)phenyl]phenyl]-N-phenylnaphthalen-1-amine
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC6=CC=CC=C65)C7=CC=CC8=CC=CC=C87

Specifications

Melting Point 279°C to 283°C
Color Yellow
Quantity 250 mg
Beilstein 8022378
Formula Weight 588.75
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N,N'-Di(1-naphthyl)-N,N'-diphenylbenzidine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only