missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di(1-naphthyl)benzidine (purified by sublimation) 98.0+%, TCI America™

Catalog No. d47681g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
D47681G 1 g
D47685G 5 g
2 options

Catalog No. D47681G

Supplier: TCI America D47681G

Only null left

Chemical Identifiers

CAS 152670-41-2
Molecular Formula C32H24N2
Molecular Weight (g/mol) 436.558
MDL Number MFCD09261380
InChI Key SZDXPEWZZGNIBB-UHFFFAOYSA-N
Synonym n,n'-di 1-naphthyl-4,4'-benzidine, n4,n4'-di naphthalen-1-yl-1,1'-biphenyl-4,4'-diamine, n4,n4'-di-naphthalen-1-yl-biphenyl-4,4'-diamine, n4,n4'-di naphthalen-1-yl biphenyl-4,4'-diamine, n,n'-di 1-naphthyl benzidine, 1,1'-biphenyl-4,4'-diamine,n4,n4'-di-1-naphthalenyl, n, n'-dinaphthol-benzidine, n4,n4'-bis naphthalen-1-yl-1,1'-biphenyl-4,4'-diamine, pubchem12682, acmc-1bwou
PubChem CID 18958605
IUPAC Name N-[4-[4-(naphthalen-1-ylamino)phenyl]phenyl]naphthalen-1-amine
SMILES C1=CC=C2C(=C1)C=CC=C2NC3=CC=C(C=C3)C4=CC=C(C=C4)NC5=CC=CC6=CC=CC=C65

Specifications

Melting Point 251°C
Quantity 1 g
Formula Weight 436.56
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Di(1-naphthyl)benzidine (purified by sublimation)