missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)phthalimide, 99%

CAS: 1515-72-6 | C12H13NO2 | 203.241 g/mol

Supplier:  Thermo Scientific Chemicals A1602014

 View more versions of this product

Catalog No. AAA1602014


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-(n-Butyl)phthalimide
32°C to 35°C
C12H13NO2
25 g
n-butylphthalimide, 2-butylisoindoline-1,3-dione, n-n-butyl phthalimide, 1-phthalimidobutane, phthalimide, n-butyl, 1h-isoindole-1,3 2h-dione, 2-butyl, unii-5th1dkt35e, 2-butyl-1h-isoindole-1,3 2h-dione, 5th1dkt35e, 2-butylbenzo c azoline-1,3-dione
CCCCN1C(=O)C2=CC=CC=C2C1=O
203.241
203.24
1515-72-6
120°C to 123°C (5 mmHg)
MFCD00039695
147784
DLKDEVCJRCPTLN-UHFFFAOYSA-N
2-butylisoindole-1,3-dione
73812
99%

Chemical Identifiers

1515-72-6
203.241
DLKDEVCJRCPTLN-UHFFFAOYSA-N
73812
CCCCN1C(=O)C2=CC=CC=C2C1=O
C12H13NO2
MFCD00039695
n-butylphthalimide, 2-butylisoindoline-1,3-dione, n-n-butyl phthalimide, 1-phthalimidobutane, phthalimide, n-butyl, 1h-isoindole-1,3 2h-dione, 2-butyl, unii-5th1dkt35e, 2-butyl-1h-isoindole-1,3 2h-dione, 5th1dkt35e, 2-butylbenzo c azoline-1,3-dione
2-butylisoindole-1,3-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-157-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only