missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)phthalimide, 99%

Catalog No. AAA1602006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1602006 5 g
AAA1602014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1602006 Supplier Thermo Scientific Chemicals Supplier No. A1602006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1515-72-6 | C12H13NO2 | 203.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1515-72-6
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00039695
InChI Key DLKDEVCJRCPTLN-UHFFFAOYSA-N
Synonym n-butylphthalimide, 2-butylisoindoline-1,3-dione, n-n-butyl phthalimide, 1-phthalimidobutane, phthalimide, n-butyl, 1h-isoindole-1,3 2h-dione, 2-butyl, unii-5th1dkt35e, 2-butyl-1h-isoindole-1,3 2h-dione, 5th1dkt35e, 2-butylbenzo c azoline-1,3-dione
PubChem CID 73812
IUPAC Name 2-butylisoindole-1,3-dione
SMILES CCCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 32°C to 35°C
Boiling Point 120°C to 123°C (5 mmHg)
Quantity 5 g
Beilstein 147784
Formula Weight 203.24
Percent Purity 99%
Chemical Name or Material N-(n-Butyl)phthalimide
EINECSNumber 216-157-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only