missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(vinylsulfonylacetyl)ethylenediamine 95.0+%, TCI America™

Catalog No. B17461G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17461G Supplier TCI America Supplier No. B17461G

Chemical Identifiers

CAS 66710-66-5
Molecular Formula C10H16N2O6S2
Molecular Weight (g/mol) 324.366
MDL Number MFCD00671499
InChI Key QWZOJDWOQYTACD-UHFFFAOYSA-N
Synonym Ethylenebis(vinylsulfonylacetamide)
PubChem CID 551576
IUPAC Name 2-ethenylsulfonyl-N-[2-[(2-ethenylsulfonylacetyl)amino]ethyl]acetamide
SMILES C=CS(=O)(=O)CC(=O)NCCNC(=O)CS(=O)(=O)C=C

Specifications

Melting Point 157°C
Color White
Quantity 1 g
Formula Weight 324.37
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(vinylsulfonylacetyl)ethylenediamine
EINECSNumber (2)-3222
TSCA No