missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Bis(trimethylsilyl)urea, 98+%
Description
N,N'-Bis(trimethylsilyl)urea is used as a reagent for the silylation of alcohols and carboxylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N′-Bis(trimethylsilyl)urea is used as a reagent for the silylation of alcohols and carboxylic acids.
Solubility
Reacts with water.
Notes
Moisture Sensitive
N,N′-Bis(trimethylsilyl)urea is used as a reagent for the silylation of alcohols and carboxylic acids.
Solubility
Reacts with water.
Notes
Moisture Sensitive
Specifications
Specifications
| Melting Point | ∽230°C (decomposition) |
| Quantity | 50 g |
| Sensitivity | Moisture sensitive |
| Beilstein | 1937452 |
| Solubility Information | Reacts with water. |
| Formula Weight | 204.42 |
| Percent Purity | ≥98% |
| Chemical Name or Material | N,N'-Bis(trimethylsilyl)urea |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 242-177-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only