missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(trimethylsilyl)urea, 98+%

Catalog No. AAL0130118
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0130118 50 g
AAL0130130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0130118 Supplier Thermo Scientific Chemicals Supplier No. L0130118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18297-63-7 | C7H20N2OSi2 | 204.42 g/mol

N,N'-Bis(trimethylsilyl)urea is used as a reagent for the silylation of alcohols and carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Bis(trimethylsilyl)urea is used as a reagent for the silylation of alcohols and carboxylic acids.

Solubility
Reacts with water.

Notes
Moisture Sensitive

Chemical Identifiers

CAS 18297-63-7
Molecular Formula C7H20N2OSi2
Molecular Weight (g/mol) 204.42
MDL Number MFCD00008260
InChI Key MASDFXZJIDNRTR-UHFFFAOYSA-N
Synonym 1,3-bis trimethylsilyl urea, n,n'-bis trimethylsilyl urea, bis trimethylsilyl urea, urea, n,n'-bis trimethylsilyl, n,n'-bis-trimethylsilyl-urea, urea, 1,3-bis trimethylsilyl, bsu, urea, bis-tms, hexamethyl disilaurea
PubChem CID 87562
IUPAC Name 1,3-bis(trimethylsilyl)urea
SMILES C[Si](C)(C)NC(=O)N[Si](C)(C)C

Specifications

Melting Point ∽230°C (decomposition)
Quantity 50 g
Sensitivity Moisture sensitive
Beilstein 1937452
Solubility Information Reacts with water.
Formula Weight 204.42
Percent Purity ≥98%
Chemical Name or Material N,N'-Bis(trimethylsilyl)urea
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 242-177-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only