missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea 98.0+%, TCI America™

Supplier:  TCI America B453525G

 View more versions of this product

Catalog No. B453525G


Only null left

Description

Description

Specifications

Specifications

123°C
107819-90-9
C12H22N2O4S
1,3-di-boc-2-methylisothiourea, s-methyl-n,n'-bis tert-butoxycarbonyl isothiourea, 1,3-bis tert-butoxycarbonyl-2-methyl-2-thiopseudourea, n,n'-di-boc-s-methylisothiourea, n,n inverted exclamation marka-di-boc-s-methylisothiourea, n,n-di-boc-s-methylisothiourea, bis-boc-thiopseudourea, polymer-bound, 1,3-di-tert-butoxycarbonyl-2-methylisothiourea, 1,3-bis-tert-butoxycarbonyl-2-methyl-thiopseudourea, z-n,n'-bis tert-butyloxycarbonyl-s-methylisothiourea
CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)SC
25 g
6376008
≥98.0% (HPLC,N)
N,N′-Bis(tert-butoxycarbonyl)-S-methylisothiourea
White
MFCD00239356
UQJXXWHAJKRDKY-UHFFFAOYSA-N
tert-butyl (NZ)-N-[[(2-methylpropan-2-yl)oxycarbonylamino]-methylsulfanylmethylidene]carbamate
290.378
290.38
Crystalline Powder

Chemical Identifiers

107819-90-9
290.378
UQJXXWHAJKRDKY-UHFFFAOYSA-N
6376008
CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)SC
C12H22N2O4S
MFCD00239356
1,3-di-boc-2-methylisothiourea, s-methyl-n,n'-bis tert-butoxycarbonyl isothiourea, 1,3-bis tert-butoxycarbonyl-2-methyl-2-thiopseudourea, n,n'-di-boc-s-methylisothiourea, n,n inverted exclamation marka-di-boc-s-methylisothiourea, n,n-di-boc-s-methylisothiourea, bis-boc-thiopseudourea, polymer-bound, 1,3-di-tert-butoxycarbonyl-2-methylisothiourea, 1,3-bis-tert-butoxycarbonyl-2-methyl-thiopseudourea, z-n,n'-bis tert-butyloxycarbonyl-s-methylisothiourea
tert-butyl (NZ)-N-[[(2-methylpropan-2-yl)oxycarbonylamino]-methylsulfanylmethylidene]carbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions