missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea 98.0+%, TCI America™

Catalog No. B45355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45355G 5 g
B453525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45355G Supplier TCI America Supplier No. B45355G

Chemical Identifiers

CAS 107819-90-9
Molecular Formula C12H22N2O4S
Molecular Weight (g/mol) 290.378
MDL Number MFCD00239356
InChI Key UQJXXWHAJKRDKY-UHFFFAOYSA-N
Synonym 1,3-di-boc-2-methylisothiourea, s-methyl-n,n'-bis tert-butoxycarbonyl isothiourea, 1,3-bis tert-butoxycarbonyl-2-methyl-2-thiopseudourea, n,n'-di-boc-s-methylisothiourea, n,n inverted exclamation marka-di-boc-s-methylisothiourea, n,n-di-boc-s-methylisothiourea, bis-boc-thiopseudourea, polymer-bound, 1,3-di-tert-butoxycarbonyl-2-methylisothiourea, 1,3-bis-tert-butoxycarbonyl-2-methyl-thiopseudourea, z-n,n'-bis tert-butyloxycarbonyl-s-methylisothiourea
PubChem CID 6376008
IUPAC Name tert-butyl (NZ)-N-[[(2-methylpropan-2-yl)oxycarbonylamino]-methylsulfanylmethylidene]carbamate
SMILES CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)SC

Specifications

Melting Point 123°C
Color White
Quantity 5 g
Formula Weight 290.38
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(tert-butoxycarbonyl)-S-methylisothiourea