missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(tert-butoxycarbonyl)-1H-pyrazole-1-carboxamidine 98.0+%, TCI America™

Catalog No. B36191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36191G 1 g
B36195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36191G Supplier TCI America Supplier No. B36191G

Chemical Identifiers

CAS 152120-54-2
Molecular Formula C14H22N4O4
Molecular Weight (g/mol) 310.354
MDL Number MFCD01075122
InChI Key QFNFDHNZVTWZED-UHFFFAOYSA-N
Synonym n,n'-di-boc-1h-pyrazole-1-carboxamidine, bis-boc-pyrazolocarboxamidine, pyrazol boc 2, n,n-bis-boc-1-guanylpyrazole, n,n'-bis-boc-1-guanylpyrazol, n,n'-bis-boc-1-guanylpyrazole, n,n'-bisboc-pyrazole-1-carboximidamide, tert-butyl tert-butoxycarbonyl amino 1h-pyrazol-1-yl methylene carbamate, tert-butyl boc-amino 1h-pyrazol-1-yl methylene carbamate, 1-n,n'-di-boc amidino pyrazole
PubChem CID 6383521
IUPAC Name tert-butyl (NZ)-N-[[(2-methylpropan-2-yl)oxycarbonylamino]-pyrazol-1-ylmethylidene]carbamate
SMILES CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)N1C=CC=N1

Specifications

Melting Point 90°C
Color White
Quantity 1 g
Formula Weight 310.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(tert-butoxycarbonyl)-1H-pyrazole-1-carboxamidine