missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Bis(salicylidene)ethylenediamine 99.0+%, TCI America™
$111.84 - $671.06
Chemical Identifiers
| CAS | 94-93-9 |
|---|---|
| Molecular Formula | C16H16N2O2 |
| Molecular Weight (g/mol) | 268.32 |
| MDL Number | MFCD00002244 |
| InChI Key | RQHVNNWVDLRULK-XSYHWHKQSA-N |
| Synonym | salen, salen unspecified, disalicylaldehyde ethylenediamine, disalicylidene-1,2-ethanediamine, n,n'-disalicylidene ethylenediamine, ethylenediamine, n,n'-disalicylidene, n,n'-bis salicylidene ethylenediamine, o-cresol, alpha,alpha-ethylenediiminodi, alpha,alpha'-ethylenedinitrilodi-o-cresol, disalicylalethylenediamine |
| PubChem CID | 5464119 |
| IUPAC Name | (6Z)-6-({[2-({[(1Z)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)ethyl]amino}methylidene)cyclohexa-2,4-dien-1-one |
| SMILES | O=C1C=CC=C\C1=C\NCCN\C=C1\C=CC=CC1=O |
Chemical Identifiers
| 94-93-9 | |
| 268.32 | |
| RQHVNNWVDLRULK-XSYHWHKQSA-N | |
| 5464119 | |
| O=C1C=CC=C\C1=C\NCCN\C=C1\C=CC=CC1=O |
| C16H16N2O2 | |
| MFCD00002244 | |
| salen, salen unspecified, disalicylaldehyde ethylenediamine, disalicylidene-1,2-ethanediamine, n,n'-disalicylidene ethylenediamine, ethylenediamine, n,n'-disalicylidene, n,n'-bis salicylidene ethylenediamine, o-cresol, alpha,alpha-ethylenediiminodi, alpha,alpha'-ethylenedinitrilodi-o-cresol, disalicylalethylenediamine | |
| (6Z)-6-({[2-({[(1Z)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)ethyl]amino}methylidene)cyclohexa-2,4-dien-1-one |
Specifications
| 94-93-9 | |
| Yellow | |
| MFCD00002244 | |
| salen, salen unspecified, disalicylaldehyde ethylenediamine, disalicylidene-1,2-ethanediamine, n,n'-disalicylidene ethylenediamine, ethylenediamine, n,n'-disalicylidene, n,n'-bis salicylidene ethylenediamine, o-cresol, alpha,alpha-ethylenediiminodi, alpha,alpha'-ethylenedinitrilodi-o-cresol, disalicylalethylenediamine | |
| O=C1C=CC=C\C1=C\NCCN\C=C1\C=CC=CC1=O | |
| 268.32 | |
| 268.32 | |
| Crystalline Powder |
| 127°C | |
| C16H16N2O2 | |
| 25 g | |
| RQHVNNWVDLRULK-XSYHWHKQSA-N | |
| (6Z)-6-({[2-({[(1Z)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)ethyl]amino}methylidene)cyclohexa-2,4-dien-1-one | |
| 5464119 | |
| ≥99.0% (T) | |
| N,N′-Bis(salicylidene)ethylenediamine |
Safety and Handling
EINECSNumber : (3)-2919
RTECSNumber : SL3780000
TSCA : Yes