missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Bis(salicylidene)-1,3-propanediamine 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 53°C |
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 282.34 |
| Percent Purity | ≥99.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N,N′-Bis(salicylidene)-1,3-propanediamine |
Safety and Handling
| TSCA | Yes |