missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(salicylidene)-1,2-phenylenediamine 96.0+%, TCI America™

Catalog No. D25725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25725G 5 g
D257225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25725G Supplier TCI America Supplier No. D25725G

Chemical Identifiers

CAS 3946-91-6
Molecular Formula C20H16N2O2
Molecular Weight (g/mol) 316.36
MDL Number MFCD00009995
InChI Key MGJWLVOAKUZEGP-UHFFFAOYSA-N
Synonym N,N′C-Bis(2-hydroxybenzylidene)-1,2-phenylenediamine, N,N′C-Disalicylal-1,2-phenylenediamine
PubChem CID 6739301
IUPAC Name 6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]anilino]methylidene]cyclohexa-2,4-dien-1-one
SMILES C1=CC=C(C(=C1)NC=C2C=CC=CC2=O)NC=C3C=CC=CC3=O

Specifications

Melting Point 165°C
Quantity 5 g
Formula Weight 316.36
Percent Purity ≥96.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(salicylidene)-1,2-phenylenediamine