missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(dimethylaminomethylene)hydrazine 98.0+%, TCI America™

Catalog No. p-7122972
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34805G 5 g
B348025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34805G Supplier TCI America Supplier No. B34805G
Only null left

Chemical Identifiers

CAS 16114-05-9
Molecular Formula C6H14N4
Molecular Weight (g/mol) 142.206
InChI Key AYTAYKXVFZPRAF-KQQUZDAGSA-N
PubChem CID 9613942
IUPAC Name N'-[(E)-dimethylaminomethylideneamino]-N,N-dimethylmethanimidamide
SMILES CN(C)C=NN=CN(C)C

Specifications

Melting Point 78°C
Color White
Quantity 5 g
Formula Weight 142.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(dimethylaminomethylene)hydrazine
TSCA No
Recommended Storage Refrigerator