missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(carbobenzoxy)-1H-pyrazole-1-carboxamidine 98.0+%, TCI America™

Catalog No. B36051G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36051G 1 g
B36055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36051G Supplier TCI America Supplier No. B36051G

Chemical Identifiers

CAS 152120-55-3
Molecular Formula C20H18N4O4
Molecular Weight (g/mol) 378.388
MDL Number MFCD02683516
InChI Key NRBUVVTTYMTSKM-UHFFFAOYSA-N
Synonym n,n'-bis-z-1-guanylpyrazole, n,n'-bis benzyloxycarbonyl-1h-pyrazole-1-carboxamidine, n,n-bisbenzyloxycarbonyl-1-guanylpyrazole, benzyl benzyloxy carbonyl amino 1h-pyrazol-1-yl methylene carbamate, pyrazole z 2, pyrazol z 2, n,n'-bs-z-1-guanylpyrazole, n,n;-bis-z-1-guanylpyrazole, benzyl n-1z-benzyloxy carbonyl imino pyrazol-1-yl methyl carbamate, n,n'-di-cbz-1h-pyrazole-1-carbo
PubChem CID 6371643
IUPAC Name benzyl (NZ)-N-[phenylmethoxycarbonylamino(pyrazol-1-yl)methylidene]carbamate
SMILES C1=CC=C(C=C1)COC(=O)NC(=NC(=O)OCC2=CC=CC=C2)N3C=CC=N3

Specifications

Melting Point 93°C
Color White
Quantity 1 g
Formula Weight 378.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(carbobenzoxy)-1H-pyrazole-1-carboxamidine