missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(9,9-dimethyl-9H-fluoren-2-yl)-N,N'-diphenylbenzidine 98.0+%, TCI America™

Catalog No. b49621g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B49621G Supplier TCI America Supplier No. B49621G

Specifications

CAS 361486-60-4
Melting Point 268°C
Molecular Formula C54H44N2
Quantity 1 g
MDL Number MFCD20528032
InChI Key VZYZZKOUCVXTOJ-UHFFFAOYSA-N
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)N(C4=CC=CC=C4)C5=CC=C(C=C5)C6=CC=C(C=C6)N(C7=CC=CC=C7)C8=CC9=C(C=C8)C1=CC=CC=C1C9(C)C)C
IUPAC Name N-[4-[4-(N-(9,9-dimethylfluoren-2-yl)anilino)phenyl]phenyl]-9,9-dimethyl-N-phenylfluoren-2-amine
Molecular Weight (g/mol) 720.96
PubChem CID 20603493
Formula Weight 720.96
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(9,9-dimethyl-9H-fluoren-2-yl)-N,N′-diphenylbenzidine
Show More Show Less