missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline 98.0+%, TCI America™

Catalog No. B5045200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5045200MG 200 mg
B50451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5045200MG Supplier TCI America Supplier No. B5045200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 165320-27-4
Molecular Formula C36H31N
Molecular Weight (g/mol) 477.651
InChI Key OZCQPISJVFUHIH-UHFFFAOYSA-N
PubChem CID 57573242
IUPAC Name N-(9,9-dimethylfluoren-2-yl)-9,9-dimethyl-N-phenylfluoren-2-amine
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)N(C4=CC=CC=C4)C5=CC6=C(C=C5)C7=CC=CC=C7C6(C)C)C

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 200 mg
Formula Weight 477.65
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline