missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(5-hydroxysalicylidene)ethylenediamine 98.0+%, TCI America™

Catalog No. p-7119893
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30151G 1 g
B30155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30151G Supplier TCI America Supplier No. B30151G
Only null left

Chemical Identifiers

CAS 124061-43-4
Molecular Formula C16H16N2O4
Molecular Weight (g/mol) 300.314
MDL Number MFCD04039747
InChI Key KNARIIFAOCMXAR-HWAYABPNSA-N
Synonym n,n'-bis 5-hydroxysalicylidene ethylenediamine, n,n'-ethylenebis 5-hydroxysalicylideneimine, n,n-bis 5-hydroxysalicylidene ethylenediamine, 2,2'-ethane-1,2-diylbis azanylylidene bis methanylylidene bis benzene-1,4-diol
PubChem CID 24985320
IUPAC Name (6Z)-4-hydroxy-6-[[2-[[(Z)-(3-hydroxy-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]ethylamino]methylidene]cyclohexa-2,4-dien-1-one
SMILES C1=CC(=O)C(=CNCCNC=C2C=C(C=CC2=O)O)C=C1O

Specifications

Melting Point 250°C
Quantity 1 g
Formula Weight 300.31
Percent Purity ≥98.0% (HPLC,N)
Physical Form Powder
Chemical Name or Material N,N′-Bis(5-hydroxysalicylidene)ethylenediamine