missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(4-methoxy-2-methylphenyl)-N,N'-diphenylbenzidine (purified by sublimation) 98.0+%, TCI America™

Catalog No. B51405G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51405G 5 g
B51401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51405G Supplier TCI America Supplier No. B51405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 169685-34-1
Molecular Formula C40H36N2O2
Molecular Weight (g/mol) 576.74
MDL Number MFCD28384132
InChI Key HACXFZALVVMGPO-UHFFFAOYSA-N
PubChem CID 23517938
IUPAC Name 4-methoxy-N-[4-[4-(N-(4-methoxy-2-methylphenyl)anilino)phenyl]phenyl]-2-methyl-N-phenylaniline
SMILES CC1=C(C=CC(=C1)OC)N(C2=CC=CC=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)N(C5=CC=CC=C5)C6=C(C=C(C=C6)OC)C

Specifications

Melting Point 160°C
Color Yellow
Quantity 5 g
Formula Weight 576.74
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(4-methoxy-2-methylphenyl)-N,N′-diphenylbenzidine (purified by sublimation)
EINECSNumber (4)-1917
TSCA No