missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(4-formylphenyl)-N,N'-diphenylbenzidine 98.0+%, TCI America™

Catalog No. b4793200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4793200MG 200 mg
B47931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4793200MG Supplier TCI America Supplier No. B4793200MG
Only null left

Chemical Identifiers

CAS 137911-28-5
Molecular Formula C38H28N2O2
Molecular Weight (g/mol) 544.654
MDL Number MFCD03093284
InChI Key DMVJDNDXIBORDT-UHFFFAOYSA-N
Synonym 4,4′C-Bis[N-(4-formylphenyl)-N-phenylamino]biphenyl
PubChem CID 17819005
IUPAC Name 4-(N-[4-[4-(N-(4-formylphenyl)anilino)phenyl]phenyl]anilino)benzaldehyde
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)C=O)C3=CC=C(C=C3)C4=CC=C(C=C4)N(C5=CC=CC=C5)C6=CC=C(C=C6)C=O

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 544.65
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(4-formylphenyl)-N,N′-diphenylbenzidine