missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-di(1-naphthyl)benzidine 98.0+%, TCI America™

Catalog No. B50931G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50931G 1 g
B50935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50931G Supplier TCI America Supplier No. B50931G
Only null left

Chemical Identifiers

CAS 910058-11-6
Molecular Formula C68H50N4
Molecular Weight (g/mol) 923.176
MDL Number MFCD20488037
InChI Key NXTRQJAJPCXJPY-UHFFFAOYSA-N
PubChem CID 22983487
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)N(C4=CC=C(C=C4)C5=CC=C(C=C5)N(C6=CC=C(C=C6)N(C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC1=CC=CC=C19)C1=CC=CC2=CC=CC=C21

Specifications

Quantity 1 g
Formula Weight 923.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis[4-(diphenylamino)phenyl]-N,N′-di(1-naphthyl)benzidine