missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(3-aminopropyl)-1,4-butanediamine Tetrahydrochloride 98.0+%, TCI America™

Catalog No. p-7119068
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14685G 5 g
B146825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14685G Supplier TCI America Supplier No. B14685G
Only null left

Chemical Identifiers

CAS 306-67-2
Molecular Formula C10H30Cl4N4
Molecular Weight (g/mol) 348.18
MDL Number MFCD00012914
InChI Key XLDKUDAXZWHPFH-UHFFFAOYSA-N
Synonym spermine tetrahydrochloride, n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride, gerontine tetrahydrochloride, neuridine tetrahydrochloride, n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride, musculamine tetrahydrochloride, geontine tetrahydrochloride, spermine hcl, 1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride, spermine, tetrahydrochloride
PubChem CID 9384
IUPAC Name tetrahydrogen (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine tetrachloride
SMILES [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN

Specifications

Melting Point 315°C
Color White
Quantity 5 g
Formula Weight 348.18
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(3-aminopropyl)-1,4-butanediamine Tetrahydrochloride
RTECSNumber EJ7230000
TSCA Yes