missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(3-aminopropyl)-1,3-propanediamine, 94%

Catalog No. AC402280050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC402280050 5 g
AC402280250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC402280050 Supplier Thermo Scientific Chemicals Supplier No. 402280050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4605-14-5 | C9H24N4 | 188.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4605-14-5
Molecular Formula C9H24N4
Molecular Weight (g/mol) 188.32
MDL Number MFCD00008213
InChI Key ZAXCZCOUDLENMH-UHFFFAOYSA-N
Synonym norspermine, n,n'-bis 3-aminopropyl-1,3-propanediamine, 1,3-propanediamine, n,n'-bis 3-aminopropyl, n,n'-bis 3-aminopropyl propane-1,3-diamine, 1,5,9,13-tetraazatridecane, 3,3,3-tetramine, thermine, 1,3-propanediamine, n1,n3-bis 3-aminopropyl, n,n-bis 3-aminopropyl-1,3-propanediamine, n1,n1'-propane-1,3-diyl bis propane-1,3-diamine
PubChem CID 78350
ChEBI CHEBI:45718
IUPAC Name (3-aminopropyl)({3-[(3-aminopropyl)amino]propyl})amine
SMILES NCCCNCCCNCCCN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.9200g/mL
Boiling Point 102°C (1.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4900 to 1.492
Linear Formula NH2(CH2)3NH(CH2)3NH(CH2)3NH2
Quantity 5 g
Specific Gravity 0.92
Formula Weight 188.32
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N, N'-Bis(3-aminopropyl)-1, 3-propanediamine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 225-007-8
TSCA TSCA

RUO – Research Use Only