missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(3,5-dimethylphenyl)-3,4,9,10-perylenetetracarboxylic Diimide 95.0+%, TCI America™

Catalog No. p-7123418
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42311G 1 g
B42315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42311G Supplier TCI America Supplier No. B42311G
Only null left

Chemical Identifiers

CAS 4948-15-6
Molecular Formula C40H26N2O4
Molecular Weight (g/mol) 598.658
MDL Number MFCD01318472
InChI Key FDXVHZCFTCIKDD-UHFFFAOYSA-N
Synonym Pigment Red 149
PubChem CID 62555
SMILES CC1=CC(=CC(=C1)N2C(=O)C3=C4C(=CC=C5C4=C(C=C3)C6=C7C5=CC=C8C7=C(C=C6)C(=O)N(C8=O)C9=CC(=CC(=C9)C)C)C2=O)C

Specifications

Quantity 1 g
Formula Weight 598.66
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(3,5-dimethylphenyl)-3,4,9,10-perylenetetracarboxylic Diimide
EINECSNumber (5)-3266&(5)-4071
TSCA Yes