missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(2-pyridylmethyl)-1,2-ethylenediamine Tetrahydrochloride Dihydrate 98.0+%, TCI America™

Catalog No. B44781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44781G 1 g
B44785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44781G Supplier TCI America Supplier No. B44781G

Chemical Identifiers

CAS 1310544-60-5
Molecular Formula C14H26Cl4N4O2
Molecular Weight (g/mol) 424.188
InChI Key OQNCOXYRPPPEIW-UHFFFAOYSA-N
PubChem CID 118705043
IUPAC Name N,N'-bis(pyridin-2-ylmethyl)ethane-1,2-diamine;dihydrate;tetrahydrochloride
SMILES C1=CC=NC(=C1)CNCCNCC2=CC=CC=N2.O.O.Cl.Cl.Cl.Cl

Specifications

Melting Point 221°C
Color White
Quantity 1 g
Formula Weight 388.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(2-pyridylmethyl)-1,2-ethylenediamine Tetrahydrochloride Dihydrate