missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%

CAS: 4439-20-7 | C6H16N2O2 | 148.2 g/mol

$158.37 - $158.37

Chemical Identifiers

CAS 4439-20-7
Molecular Formula C6H16N2O2
Molecular Weight (g/mol) 148.2
MDL Number MFCD00002842
InChI Key GFIWSSUBVYLTRF-UHFFFAOYSA-N
Synonym n,n'-bis 2-hydroxyethyl ethylenediamine, n,n'-diethanolethylenediamine, ethanol, 2,2'-1,2-ethanediyldiimino bis, 1,2-bis 2-hydroxyethyl amino ethane, 3,6-diazaoctane-1,8-diol, 2,2'-ethane-1,2-diylbis azanediyl diethanol, n,n'-bis 2-hydroxyethyl ethylene diamine, n,n'-bis 2-hydroxyethyl-ethylenediamine, 2-2-2-hydroxyethylamino ethylamino ethanol
PubChem CID 78179
IUPAC Name 2-[2-(2-hydroxyethylamino)ethylamino]ethanol
SMILES C(CNCCO)NCCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC345180100
SDS View Documents
Thermo Scientific Chemicals
345180100
10 g Glass bottle
Each for $158.37
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4439-20-7
148.2
GFIWSSUBVYLTRF-UHFFFAOYSA-N
78179
C(CNCCO)NCCO
C6H16N2O2
MFCD00002842
n,n'-bis 2-hydroxyethyl ethylenediamine, n,n'-diethanolethylenediamine, ethanol, 2,2'-1,2-ethanediyldiimino bis, 1,2-bis 2-hydroxyethyl amino ethane, 3,6-diazaoctane-1,8-diol, 2,2'-ethane-1,2-diylbis azanediyl diethanol, n,n'-bis 2-hydroxyethyl ethylene diamine, n,n'-bis 2-hydroxyethyl-ethylenediamine, 2-2-2-hydroxyethylamino ethylamino ethanol
2-[2-(2-hydroxyethylamino)ethylamino]ethanol

Specifications

4439-20-7
100.0
White to Yellow
95%
C6H16N2O2
MFCD00002842
n,n'-bis 2-hydroxyethyl ethylenediamine, n,n'-diethanolethylenediamine, ethanol, 2,2'-1,2-ethanediyldiimino bis, 1,2-bis 2-hydroxyethyl amino ethane, 3,6-diazaoctane-1,8-diol, 2,2'-ethane-1,2-diylbis azanediyl diethanol, n,n'-bis 2-hydroxyethyl ethylene diamine, n,n'-bis 2-hydroxyethyl-ethylenediamine, 2-2-2-hydroxyethylamino ethylamino ethanol
C(CNCCO)NCCO
148.2
148.2
Chunks and/or Powder
94.0
92°C to 99°C
Authentic
Glass bottle
(HOCH2CH2NHCH2-)2
10 g
GFIWSSUBVYLTRF-UHFFFAOYSA-N
2-[2-(2-hydroxyethylamino)ethylamino]ethanol
78179
95%
N, N'-Bis(2-hydroxyethyl)ethylenediamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-656-4

SDS
Documents

Documents

RUO – Research Use Only