missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(2-cyanoethyl)formamide 90.0+%, TCI America™

Catalog No. B09775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09775G 5 g
B097725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09775G Supplier TCI America Supplier No. B09775G

Chemical Identifiers

CAS 3445-84-9
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.17
MDL Number MFCD00040894
InChI Key MYRFNYCEQURXPT-UHFFFAOYSA-N
Synonym n,n-bis 2-cyanoethyl formamide, formamide, n,n-bis 2-cyanoethyl, 3,3'-formyliminodipropiononitrile, 3,3'-formylimino dipropionitrile, luprintan, acmc-1cm2w, bis 2-cyanoethyl formamide, bis 2-cyanoethyl formamide #, nn-bis 2-cyanoethyl formamide, formamide, bis 2-cyanoethyl
PubChem CID 76983
IUPAC Name N,N-bis(2-cyanoethyl)formamide
SMILES O=CN(CCC#N)CCC#N

Specifications

Boiling Point 122°C
Quantity 5 g
UN Number 2810
Formula Weight 151.17
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Bis(2-cyanoethyl)formamide
EINECSNumber (2)-1529&(7)-0340
TSCA Yes