missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic Diimide 98.0+%, TCI America™
Supplier: TCI America B42685G
Specifications
| N,N′-Bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic Diimide | |
| C48H42N2O4 | |
| Perylene Orange | |
| CC(C)C1=C(C(=CC=C1)C(C)C)N2C(=O)C3=C4C(=CC=C5C4=C(C=C3)C6=C7C5=CC=C8C7=C(C=C6)C(=O)N(C8=O)C9=C(C=CC=C9C(C)C)C(C)C)C2=O | |
| 14618068 | |
| ≥98.0% (N) |
| 82953-57-9 | |
| 5 g | |
| NAZODJSYHDYJGP-UHFFFAOYSA-N | |
| 710.874 | |
| 710.87 | |
| Crystalline Powder |
Chemical Identifiers
| 82953-57-9 | |
| 710.874 | |
| Perylene Orange | |
| CC(C)C1=C(C(=CC=C1)C(C)C)N2C(=O)C3=C4C(=CC=C5C4=C(C=C3)C6=C7C5=CC=C8C7=C(C=C6)C(=O)N(C8=O)C9=C(C=CC=C9C(C)C)C(C)C)C2=O |
| C48H42N2O4 | |
| NAZODJSYHDYJGP-UHFFFAOYSA-N | |
| 14618068 |
Safety and Handling
TSCA : No