missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic Diimide 98.0+%, TCI America™

Catalog No. B42681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42681G 1 g
B42685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42681G Supplier TCI America Supplier No. B42681G

Chemical Identifiers

CAS 82953-57-9
Molecular Formula C48H42N2O4
Molecular Weight (g/mol) 710.874
InChI Key NAZODJSYHDYJGP-UHFFFAOYSA-N
Synonym Perylene Orange
PubChem CID 14618068
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2C(=O)C3=C4C(=CC=C5C4=C(C=C3)C6=C7C5=CC=C8C7=C(C=C6)C(=O)N(C8=O)C9=C(C=CC=C9C(C)C)C(C)C)C2=O

Specifications

Quantity 1 g
Formula Weight 710.87
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic Diimide