missing translation for 'onlineSavingsMsg'
Learn More

N,N'-1,4-Phenylenedimaleimide 98.0+%, TCI America™

Catalog No. P07781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07781G 1 g
P07785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P07781G Supplier TCI America Supplier No. P07781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3278-31-7
Molecular Formula C14H8N2O4
Molecular Weight (g/mol) 268.23
MDL Number MFCD00005505
InChI Key AQGZJQNZNONGKY-UHFFFAOYSA-N
Synonym n,n'-1,4-phenylenedimaleimide, n,n'-p-phenylenedimaleimide, n,n'-4-phenylenedimaleimide, phenyl-1,4-bismaleimide, unii-bec7p1e6j1, 1,1'-1,4-phenylene bis 1h-pyrrole-2,5-dione, bec7p1e6j1, 1-4-2,5-dioxopyrrol-1-yl phenyl pyrrole-2,5-dione, p-pdm, 1,4-dimaleimidobenzene
PubChem CID 76765
IUPAC Name 1-[4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)phenyl]-2,5-dihydro-1H-pyrrole-2,5-dione
SMILES O=C1C=CC(=O)N1C1=CC=C(C=C1)N1C(=O)C=CC1=O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 268.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-1,4-Phenylenedimaleimide