missing translation for 'onlineSavingsMsg'
Learn More

N-Methylsuccinimide 98.0+%, TCI America™

Catalog No. p-7133091
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22075G 5 g
M220725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22075G Supplier TCI America Supplier No. M22075G

Chemical Identifiers

CAS 1121-07-9
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00005517
InChI Key KYEACNNYFNZCST-UHFFFAOYSA-N
Synonym n-methylsuccinimide, 2,5-pyrrolidinedione, 1-methyl, succinimide, n-methyl, 1-methyl-2,5-pyrrolidinedione, n-methyl succinimide, acmc-2099cm, 1-methylazolidine-2,5-dione, ksc492s6n, 1-methyl-pyrrolidine-2,5-dione
PubChem CID 70717
IUPAC Name 1-methylpyrrolidine-2,5-dione
SMILES CN1C(=O)CCC1=O

Specifications

Melting Point 70°C
Color White
Boiling Point 235°C
Quantity 5 g
Formula Weight 113.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Methylsuccinimide
RTECSNumber UY1028650
TSCA Yes