missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyrrole-3-boronic acid pinacol ester, 95%

Catalog No. AC441000050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC441000050 5 g Glass bottle
AC441000010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC441000050 Supplier Thermo Scientific Chemicals Supplier No. 441000050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 953040-54-5 | C11H18BNO2 | 207.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 953040-54-5
Molecular Formula C11H18BNO2
Molecular Weight (g/mol) 207.08
MDL Number MFCD13182200
InChI Key SYAMJGBBLFTQTK-UHFFFAOYSA-N
Synonym 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, 1-methylpyrrole-3-boronic acid, pinacol ester, 1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, 1-methyl-1h-pyrrol-3-ylboronic acid pinacol ester, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole, pubchem18847, 1-methyl-1h-pyrrole-3-boronic acid pinacol ester, n-methylpyrrole-3-boronic acid pinacol ester, 1-methyl-pyrrol-3-ylboronic acid pinacol ester, 1-methyl-1h-pyrrol-3-yl boronic acid pinacol ester
PubChem CID 53217148
IUPAC Name 1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole
SMILES CN1C=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 44.0°C to 51.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 207.08
Percent Purity 90%
Physical Form Powder
Chemical Name or Material N-Methylpyrrole-3-boronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only