missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyrrole-2-boronic acid pinacol ester, 97%, Thermo Scientific™

$204.97

Chemical Identifiers

CAS 850567-47-4
Molecular Formula C11H18BNO2
Molecular Weight (g/mol) 207.08
InChI Key OEQQEXKRORJZPR-UHFFFAOYSA-N
Synonym 1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, 1-methyl-1h-pyrrole-2-boronic acid, pinacol ester, 1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole, 1-methyl-2-pyrroleboronic acid pinacol ester, n-methylpyrrole-2-boronic acid, pinacol ester, 1h-pyrrole, 1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem18528, 1-methylpyrrole-2-boronic acid pinacol ester, n-methyl-2-pyrroleboronic acid, pinacol ester, 1-methyl-2-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole
PubChem CID 44118773
IUPAC Name 1-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC441040010
View Documents Promotion Details
N/A 1g
Each for $204.97
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

850567-47-4
207.08
1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, 1-methyl-1h-pyrrole-2-boronic acid, pinacol ester, 1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole, 1-methyl-2-pyrroleboronic acid pinacol ester, n-methylpyrrole-2-boronic acid, pinacol ester, 1h-pyrrole, 1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem18528, 1-methylpyrrole-2-boronic acid pinacol ester, n-methyl-2-pyrroleboronic acid, pinacol ester, 1-methyl-2-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole
1-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole
C11H18BNO2
OEQQEXKRORJZPR-UHFFFAOYSA-N
44118773
B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C

Specifications

850567-47-4
100.0
96% min. (GC)
C11H18BNO2
OEQQEXKRORJZPR-UHFFFAOYSA-N
1-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole
44118773
97%
96.0
Authentic
Glass Bottle
1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, 1-methyl-1h-pyrrole-2-boronic acid, pinacol ester, 1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole, 1-methyl-2-pyrroleboronic acid pinacol ester, n-methylpyrrole-2-boronic acid, pinacol ester, 1h-pyrrole, 1-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem18528, 1-methylpyrrole-2-boronic acid pinacol ester, n-methyl-2-pyrroleboronic acid, pinacol ester, 1-methyl-2-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole
B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C
207.08
207.08
N-Methylpyrrole-2-boronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only