missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate, 97%

Catalog No. AAL1207206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1207206 5 g
AAL1207214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1207206 Supplier Thermo Scientific Chemicals Supplier No. L1207206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 82228-89-5 | C13H13NO4S | 279.31 g/mol

N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate is a useful reagent for the chemical transformation of primary amines to aldehydes and ketones under extremely mild conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate is a useful reagent for the chemical transformation of primary amines to aldehydes and ketones under extremely mild conditions.

Solubility
Sparingly soluble in water (0.27 g/L at 25°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Store under inert gas. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 82228-89-5
Molecular Formula C13H13NO4S
Molecular Weight (g/mol) 279.31
MDL Number MFCD00043164
InChI Key HSVLGIFAXFDLMU-UHFFFAOYSA-M
Synonym 4-formyl-1-methylpyridinium benzenesulfonate, 4-formyl-1-methyl-pyridinium benzenesulfonate, 4-formyl-1-methylpyridin-1-ium benzenesulfonate, pyridinium, 4-formyl-1-methyl-, benzenesulfonate, benzenesulfonate; 1-methylpyridin-1-ium-4-carbaldehyde, 4-formyl-1-methyl pyridinium benzensulfonate, 4-formyl-l-methyl pyridinium benzensulfonate, 4-formyl-1-methylpyridinium benzenesulfo nate, benzenesulfonic acid, 1-methylpyridine-4-carbaldehyde
PubChem CID 9817127
IUPAC Name benzenesulfonate;1-methylpyridin-1-ium-4-carbaldehyde
SMILES C[N+]1=CC=C(C=O)C=C1.[O-]S(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 98°C to 100°C
Quantity 5 g
Beilstein 5695963
Sensitivity Air Sensitive
Solubility Information Sparingly soluble in water (0.27g/L at 25°C).
Formula Weight 279.32 (Anhydrous)
Percent Purity 97%
Chemical Name or Material N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only