missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpropylamine 98.0+%, TCI America™

Catalog No. M22325G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22325G Supplier TCI America Supplier No. M22325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 627-35-0
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00009361
InChI Key GVWISOJSERXQBM-UHFFFAOYSA-N
Synonym n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
PubChem CID 12315
IUPAC Name N-methylpropan-1-amine
SMILES CCCNC

Specifications

Color Colorless
Boiling Point 63°C
Quantity 5 g
UN Number 2733
Formula Weight 73.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methylpropylamine