missing translation for 'onlineSavingsMsg'
Learn More

N-Methylphthalimide 99.0+%, TCI America™

Catalog No. M029925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M029925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M029925G Supplier TCI America Supplier No. M029925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 550-44-7
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00023063
InChI Key ZXLYYQUMYFHCLQ-UHFFFAOYSA-N
Synonym n-methylphthalimide, 2-methylisoindoline-1,3-dione, phthalimide, n-methyl, 1h-isoindole-1,3 2h-dione, 2-methyl, unii-647up45j2u, 2-methyl-1h-isoindole-1,3 2h-dione, n-methyl phthalimide, dsstox_cid_7198, acmc-1b11u
PubChem CID 11074
IUPAC Name 2-methylisoindole-1,3-dione
SMILES CN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 135°C
Color White
Quantity 25 g
Formula Weight 161.16
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Methylphthalimide
RTECSNumber TI5602700
TSCA Yes