missing translation for 'onlineSavingsMsg'
Learn More

N-Methylphthalimide, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 305100250

 View more versions of this product

Catalog No. AC305100250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

N-Methylphthalimide
97.5% min. (HPLC)
C9H7NO2
n-methylphthalimide, 2-methylisoindoline-1,3-dione, phthalimide, n-methyl, 1h-isoindole-1,3 2h-dione, 2-methyl, unii-647up45j2u, 2-methyl-1h-isoindole-1,3 2h-dione, n-methyl phthalimide, dsstox_cid_7198, acmc-1b11u
CN1C(=O)C2=CC=CC=C2C1=O
161.16
161.16
550-44-7
Glass bottle
25g
ZXLYYQUMYFHCLQ-UHFFFAOYSA-N
2-methylisoindole-1,3-dione
11074
98%

Chemical Identifiers

550-44-7
161.16
n-methylphthalimide, 2-methylisoindoline-1,3-dione, phthalimide, n-methyl, 1h-isoindole-1,3 2h-dione, 2-methyl, unii-647up45j2u, 2-methyl-1h-isoindole-1,3 2h-dione, n-methyl phthalimide, dsstox_cid_7198, acmc-1b11u
2-methylisoindole-1,3-dione
C9H7NO2
ZXLYYQUMYFHCLQ-UHFFFAOYSA-N
11074
CN1C(=O)C2=CC=CC=C2C1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 208-982-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only