missing translation for 'onlineSavingsMsg'
Learn More

N-Methylphthalimide, 98%

Catalog No. AC305100050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC305100050 5 g Glass bottle
AC305100250 25 g Glass bottle
AC305101000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC305100050 Supplier Thermo Scientific Chemicals Supplier No. 305100050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 550-44-7 | C9H7NO2 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 550-44-7
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
InChI Key ZXLYYQUMYFHCLQ-UHFFFAOYSA-N
Synonym n-methylphthalimide, 2-methylisoindoline-1,3-dione, phthalimide, n-methyl, 1h-isoindole-1,3 2h-dione, 2-methyl, unii-647up45j2u, 2-methyl-1h-isoindole-1,3 2h-dione, n-methyl phthalimide, dsstox_cid_7198, acmc-1b11u
PubChem CID 11074
IUPAC Name 2-methylisoindole-1,3-dione
SMILES CN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 129.0°C to 132.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material N-Methylphthalimide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 208-982-4

RUO – Research Use Only