missing translation for 'onlineSavingsMsg'
Learn More

N-Methylphthalimide, 98%, Thermo Scientific Chemicals

Catalog No. AAL0899114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0899114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0899114 Supplier Thermo Scientific Chemicals Supplier No. L0899114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 550-44-7 | Molecular Formula: C9H7NO2 | Molecular Weight: 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 550-44-7
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00023063
InChI Key ZXLYYQUMYFHCLQ-UHFFFAOYSA-N
Synonym n-methylphthalimide, 2-methylisoindoline-1,3-dione, phthalimide, n-methyl, 1h-isoindole-1,3 2h-dione, 2-methyl, unii-647up45j2u, 2-methyl-1h-isoindole-1,3 2h-dione, n-methyl phthalimide, dsstox_cid_7198, acmc-1b11u
PubChem CID 11074
IUPAC Name 2-methylisoindole-1,3-dione
SMILES CN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 132°C to 136°C
Quantity 25 g
Beilstein 124428
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material N-Methylphthalimide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-982-4
RTECSNumber TI5602700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only