missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpentylamine 98.0+%, TCI America™

Catalog No. M22311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22311G 1 g
M22315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22311G Supplier TCI America Supplier No. M22311G

Chemical Identifiers

CAS 25419-06-1
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00041354
InChI Key UOIWOHLIGKIYFE-UHFFFAOYSA-N
Synonym n-methylpentylamine, n-methylamylamine, amylmethylamine, methyl pentyl amine, 1-pentanamine, n-methyl, methylpentylamine, pentylamine, n-methyl, methyl-pentyl-amine, n-methyl-n-pentylamine, methylamylamine
PubChem CID 117479
IUPAC Name methyl(pentyl)amine
SMILES CCCCCNC

Specifications

Color Yellow
Boiling Point 118°C
Quantity 1 g
UN Number 2733
Formula Weight 101.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Methylpentylamine