missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpentylamine, 98%

Catalog No. AAL0488209
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0488209 10 g
AAL0488204 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0488209 Supplier Thermo Scientific Chemicals Supplier No. L0488209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 25419-06-1 | C6H15N | 101.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25419-06-1
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00041354
InChI Key UOIWOHLIGKIYFE-UHFFFAOYSA-N
Synonym n-methylpentylamine, n-methylamylamine, amylmethylamine, methyl pentyl amine, 1-pentanamine, n-methyl, methylpentylamine, pentylamine, n-methyl, methyl-pentyl-amine, n-methyl-n-pentylamine, methylamylamine
PubChem CID 117479
IUPAC Name N-methylpentan-1-amine
SMILES CCCCCNC

Specifications

Density 0.738
Boiling Point 117°C to 119°C
Flash Point 18°C (64°F)
Refractive Index 1.4105
Quantity 10 g
UN Number UN2733
Beilstein 1731333
Sensitivity Air Sensitive
Formula Weight 101.19
Percent Purity 98%
Chemical Name or Material N-Methylpentylamine
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 246-966-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only