missing translation for 'onlineSavingsMsg'
Learn More

n-Methyloctylamine, 98%

CAS: 2439-54-5 | C9H21N | 143.27 g/mol

$312.33 - $312.33

Chemical Identifiers

CAS 2439-54-5
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.27
MDL Number MFCD00048927
InChI Key SEGJNMCIMOLEDM-UHFFFAOYSA-N
Synonym n-methyloctylamine, methyl octyl amine, n-methyl-n-octylamine, 1-octanamine, n-methyl, octylmethylamine, methyloctylamine, unii-4o0o17jz7d, n-methyl octylamine, n-methyl-octylamine, methyl n-octyl amine
PubChem CID 75538
IUPAC Name N-methyloctan-1-amine
SMILES CCCCCCCCNC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC325620250
SDS View Documents
Thermo Scientific Chemicals
325620250
25 g Glass bottle
Each for $312.33
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2439-54-5
143.27
SEGJNMCIMOLEDM-UHFFFAOYSA-N
75538
CCCCCCCCNC
C9H21N
MFCD00048927
n-methyloctylamine, methyl octyl amine, n-methyl-n-octylamine, 1-octanamine, n-methyl, octylmethylamine, methyloctylamine, unii-4o0o17jz7d, n-methyl octylamine, n-methyl-octylamine, methyl n-octyl amine
N-methyloctan-1-amine

Specifications

2439-54-5
100.0
0.7900g/mL
63°C
97.5% min. (GC)
C9H21N
CH3(CH2)7NHCH3
25 g
n-methyloctylamine, methyl octyl amine, n-methyl-n-octylamine, 1-octanamine, n-methyl, octylmethylamine, methyloctylamine, unii-4o0o17jz7d, n-methyl octylamine, n-methyl-octylamine, methyl n-octyl amine
CCCCCCCCNC
143.27
143.27
Liquid
97.5
Colorless to Yellow
68.0°C (8.0 mmHg)
Authentic
Glass bottle
1.4260 to 1.4280
MFCD00048927
0.79
SEGJNMCIMOLEDM-UHFFFAOYSA-N
N-methyloctan-1-amine
75538
98%
n-Methyloctylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 219-462-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only