missing translation for 'onlineSavingsMsg'
Learn More

N-Methylnicotinamide 98.0+%, TCI America™

Catalog No. M037410G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M037410G 10 g
M037425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M037410G Supplier TCI America Supplier No. M037410G

Chemical Identifiers

CAS 114-33-0
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00006384
InChI Key ZYVXHFWBYUDDBM-UHFFFAOYSA-N
PubChem CID 64950
ChEBI CHEBI:64399
IUPAC Name N-methylpyridine-3-carboxamide
SMILES CNC(=O)C1=CN=CC=C1

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 10 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methylnicotinamide
EINECSNumber (9)-0954
TSCA No