missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methylmethacrylamide (stabilized with HQ) 98.0+%, TCI America™
$113.59 - $344.61
Chemical Identifiers
| CAS | 3887-02-3 |
|---|---|
| Molecular Formula | C5H9NO |
| Molecular Weight (g/mol) | 99.13 |
| MDL Number | MFCD00048115 |
| InChI Key | WFKDPJRCBCBQNT-UHFFFAOYSA-N |
| PubChem CID | 77495 |
| IUPAC Name | N,2-dimethylprop-2-enamide |
| SMILES | CC(=C)C(=O)NC |
Chemical Identifiers
| 3887-02-3 | |
| 99.13 | |
| WFKDPJRCBCBQNT-UHFFFAOYSA-N | |
| N,2-dimethylprop-2-enamide |
| C5H9NO | |
| MFCD00048115 | |
| 77495 | |
| CC(=C)C(=O)NC |
Specifications
| 3887-02-3 | |
| 110°C | |
| MFCD00048115 | |
| WFKDPJRCBCBQNT-UHFFFAOYSA-N | |
| N,2-dimethylprop-2-enamide | |
| 77495 | |
| ≥98.0% (T) | |
| N-Methylmethacrylamide (stabilized with HQ) |
| Yellow | |
| C5H9NO | |
| 5 mL | |
| CC(=C)C(=O)NC | |
| 99.13 | |
| 99.13 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator